missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds with Polyvinyl-lined Caps
Click to view available options
Capacity:
1 oz., 30mL
16 oz., 500 mL
2 oz., 60 mL
32 oz., 1000 mL
4 oz., 125 mL
8 oz., 250 mL
Packaging:
Convenience pack
Valu-bulk Pk.
Valu-bulk pack
Description
Amber Type III soda lime glass protects light-sensitive solutions
- Available in partitioned Convenience Pack or Valu-Bulk™ cartons
- Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Each case of Convenience Pack bottles includes Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
USP Type III soda-lime glass
Specifications
Specifications
Capacity | 32 oz., 1000 mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | Amber |
Diameter (English) | 3.7 in. |
Material | Soda Lime Glass |
Packaging | Valu-bulk pack |
Closure Color | White |
Height (Metric) | 208 mm |
Height (English) | 8.18 in. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'