missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Nuclepore Track-Etched Polycarbonate AOX Membranes

GE’s Nuclepore filters are manufactured from high-quality polycarbonate film, with sharply defined pore sizes and high flow rates for reproducible AOX analysis.

141.83 € - 193.00 €

Specifications

Autoclavable Autoclavable
Material Nucleopore Polycarbonate
Pore Size 0.4 μm
Flow Rate High
For Use With (Application) Epifluorescence microscopy, environmental analysis, cell biology
View More Specs

Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
11338994
View Documents Promotion Details
Cytiva
110637
25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11322905
View Documents Promotion Details
Cytiva
111137
47 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • High chemical resistance and thermal stability for a wide range of samples
  • Low, consistent ash levels and low tare weights
  • Compatible with ISO 9562 guidelines for determining adsorbable organically bound halogens
  • Smooth flat surface increases particle visibility
Specifications

Specifications

Autoclavable
0.4 μm
Epifluorescence microscopy, environmental analysis, cell biology
Nucleopore Polycarbonate
High
Hydrophilic
Videos
SDS
Documents

Documents

Product Certifications