Learn More
Cytiva Whatman™ Nuclepore Track-Etched Polycarbonate AOX Membranes
GE’s Nuclepore filters are manufactured from high-quality polycarbonate film, with sharply defined pore sizes and high flow rates for reproducible AOX analysis.
Brand: Cytiva 110637
This item is not returnable.
View return policy
Description
- High chemical resistance and thermal stability for a wide range of samples
- Low, consistent ash levels and low tare weights
- Compatible with ISO 9562 guidelines for determining adsorbable organically bound halogens
- Smooth flat surface increases particle visibility
Specifications
Autoclavable | |
0.4 μm | |
Epifluorescence microscopy, environmental analysis, cell biology | |
25 mm |
Nucleopore Polycarbonate | |
High | |
Hydrophilic |