missing translation for 'onlineSavingsMsg'
Learn More

VEGFC Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen VEGFC
Concentration 0.33 mg/mL
Applications Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16832005
View Documents
Proteintech
22601-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16822005
View Documents
Proteintech
22601-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.
Specifications

Specifications

VEGFC
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
Rabbit
Mouse, Human
P49767, P97953
22341, 7424
VEGFC Fusion Protein Ag18182
Primary
-20°C
VEGFC
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Flt4 L, Flt4 ligand, VEGF C, VEGFC, VRP
VEGFC
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.