missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22601-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.Specifications
VEGFC | |
Polyclonal | |
Unconjugated | |
VEGFC | |
Flt4 L, Flt4 ligand, VEGF C, VEGFC, VRP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22341, 7424 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49767, P97953 | |
VEGFC | |
VEGFC Fusion Protein Ag18182 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction