missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Universal Platform
OHAUS™ Universal Platform designed to hold samples and laboratory glassware.
Brand: OHAUS™ 30400057
995.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
- Platforms can increase the amount of usable space for samples
- In some cases can be stacked to increase throughput and double sample capacity
- Tube racks and dimpled mats securely hold tubes in place
Specifications
Platform | |
24.00 in. | |
61 x 61 cm | |
61.00 cm | |
61.00 cm | |
Universal |
Stainless Steel | |
61.00 cm | |
24.00 in. | |
24.00 in. | |
Universal Platform, 61 X 61 cm, S/S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction