missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Universal Platform

OHAUS™ Universal Platform designed to hold samples and laboratory glassware.

Brand:  OHAUS™ 30400056

415.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15782469

  • 426.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
  • Tube racks and dimpled mats securely hold tubes in place
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Platform
24.00 in.
61 x 45.7 cm
61.00 cm
45.70 cm
Universal
Stainless Steel
61.00 cm
24.00 in.
18.00 in.
Universal Platform, 61 X 46 cm, S/S
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.