Learn More
Uvex™ U-Fit Disposable NBR Protective Gloves
Thin, reliable NBR gloves feature a roughed glove surface for enhanced grip capabilities and provide reliable splash protection when handling chemicals such as acids, alkalis, solids, and aqueous salt solutions.
22.80 € - 26.30 €
Specifications
Color | Blue |
---|---|
Latex Free | Yes |
Material | NBR |
Palm Thickness (Metric) | 0.1 mm |
EN ISO 374-1 | Type B |
Description
- Thin, reliable NBR gloves enhanced grip capabilities due to the roughened palm surface
- Splash protection when handling chemicals such as acids, alkalis, solids, or aqueous salt solutions
- Silicone-free according to the impression test
- Highly flexible with good tactile sensitivity
- Very good mechanical strength
Specifications
Blue | |
NBR | |
Type B | |
Bacteria, Fungi WITH Virus Protection |
Yes | |
0.1 mm | |
KPT |