missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Chemo Gown
Collared gown with hook and loop closure in the neck, serged seams, knit cuffs and bound ties at waist originating from elasticated sides. Not clean-processed and not sterilized.
256.00 € - 265.00 €
Specifications
Product Type | Controlled Environments Gown |
---|---|
Material | Polyethylene |
Color | White |
Closure Type | Hook and Loop |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17876865
|
DuPont™
D15542351 |
X-Small |
256.00 €
Pack of 30 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
17856865
|
DuPont™
D15542352 |
Small |
256.00 €
Pack of 30 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
17866865
|
DuPont™
D15542353 |
Medium |
256.00 €
Pack of 30 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17836865
|
DuPont™
D15542354 |
Large |
256.00 €
Pack of 30 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
17846865
|
DuPont™
D15542355 |
X-Large |
256.00 €
Pack of 30 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
17826865
|
DuPont™
D15542356 |
2X-Large |
256.00 €
Pack of 30 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
17886865
|
DuPont™
D15542357 |
3X-Large |
265.00 €
Pack of 30 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
Description
- Available in sizes XS to 3X
- Increased skirt length ensures protection against frontal exposure Openness in the back closure increases wearer comfort
- Delivers an ideal balance of protection, durability and comfort
- Made of high density polyethylene using a patented flash spinning process
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based liquid splash
- Manufactured in a controlled environment
Specifications
Controlled Environments Gown | |
White | |
Single-use | |
Knit | |
Long Sleeve |
Polyethylene | |
Hook and Loop | |
Non-sterile | |
Yes | |
Autoclavable |