Learn More
DuPont™ Tyvek™ IsoClean™ Chemo Gown
Product Code. 17846865
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Collared gown with hook and loop closure in the neck, serged seams, knit cuffs and bound ties at waist originating from elasticated sides. Not clean-processed and not sterilized.
  • Available in sizes XS to 3X
  • Increased skirt length ensures protection against frontal exposure Openness in the back closure increases wearer comfort
  • Delivers an ideal balance of protection, durability and comfort
  • Made of high density polyethylene using a patented flash spinning process
  • Provides an inherent barrier to particles, microorganisms and non-hazardous water-based liquid splash
  • Manufactured in a controlled environment
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Product Type Controlled Environments Gown
Material Polyethylene
Closure Type Hook and Loop
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Latex Free Yes
Sleeve Style Long Sleeve
Autoclavable Autoclavable