Learn More
Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, ultrapure, 99.9%
116.58 € - 508.53 €
Chemical Identifiers
CAS | 77-86-1 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
MDL Number | MFCD00004679 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
Synonym | TRIS |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446989
|
Thermo Scientific Chemicals
J65594.A1 |
1 kg |
116.58 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15456989
|
Thermo Scientific Chemicals
J65594.A7 |
5 kg |
508.53 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis. It acts as a precursor for the preparation of polymers, oxazolones and oxazolidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis. It acts as a precursor for the preparation of polymers, oxazolones and oxazolidines.
Solubility
Soluble in water, ethylene glycol, methanol, ethanol, dimethyl formamide, acetone, ethyl acetate, olive oil, chloroform and alkaline solution.
Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
Chemical Identifiers
77-86-1 | |
121.136 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
C4H11NO3 | |
MFCD00004679 | |
TRIS | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
Specifications
77-86-1 | |
219°C to 220°C | |
1.35 | |
C4H11NO3 | |
741883 | |
TRIS | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.14 | |
Tris(hydroxymethyl)aminomethane |
167°C to 172°C | |
White | |
Crystalline Powder | |
MFCD00004679 | |
149772 | |
Soluble in water,ethylene glycol,methanol,ethanol,dimethyl formamide,acetone,ethyl acetate,olive oil,chloroform and alkaline solution. | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
99.9% | |
Ultrapure |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Warning
missing translation for 'einecsNumber' : 201-064-4
missing translation for 'rtecsNumber' : TY2900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only