Learn More
Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, ultrapure, 99.9%
Description
Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis. It acts as a precursor for the preparation of polymers, oxazolones and oxazolidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis. It acts as a precursor for the preparation of polymers, oxazolones and oxazolidines.
Solubility
Soluble in water, ethylene glycol, methanol, ethanol, dimethyl formamide, acetone, ethyl acetate, olive oil, chloroform and alkaline solution.
Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
Specifications
Specifications
Melting Point | 167°C to 172°C |
Boiling Point | 219°C to 220°C |
Color | White |
Density | 1.35 |
Physical Form | Crystalline Powder |
Quantity | 5 kg |
Beilstein | 741883 |
Merck Index | 149772 |
Solubility Information | Soluble in water,ethylene glycol,methanol,ethanol,dimethyl formamide,acetone,ethyl acetate,olive oil,chloroform and alkaline solution. |
Formula Weight | 121.14 |
Show More |
RUO – Research Use Only