Learn More
Tris(4-aminophenyl)amine, 97%, Thermo Scientific Chemicals
42.90 € - 471.00 €
Chemical Identifiers
CAS | 5981-09-9 |
---|---|
Molecular Formula | C18H18N4 |
Molecular Weight (g/mol) | 290.37 |
MDL Number | MFCD00778301 |
InChI Key | SNLFYGIUTYKKOE-UHFFFAOYSA-N |
Synonym | tris 4-aminophenyl amine, 4,4',4-triaminotriphenylamine, n1,n1-bis 4-aminophenyl benzene-1,4-diamine, 1,4-benzenediamine, n,n-bis 4-aminophenyl, n,n-bis 4-aminophenyl-1,4-benzenediamine, 1,4-benzenediamine, n1,n1-bis 4-aminophenyl, 1-n,1-n-bis 4-aminophenyl benzene-1,4-diamine, pubchem22505, tri 4-aminophenyl amine, tris 4-aminophenyl-amine |
PubChem CID | 80083 |
IUPAC Name | 4-N,4-N-bis(4-aminophenyl)benzene-1,4-diamine |
SMILES | C1=CC(=CC=C1N)N(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15480959
|
Thermo Scientific Alfa Aesar
H63963.MD |
250 mg |
42.90 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15460959
|
Thermo Scientific Alfa Aesar
H63963.03 |
1 g |
121.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15470959
|
Thermo Scientific Alfa Aesar
H63963.06 |
5 g |
471.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
5981-09-9 | |
290.37 | |
SNLFYGIUTYKKOE-UHFFFAOYSA-N | |
80083 | |
C1=CC(=CC=C1N)N(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N |
C18H18N4 | |
MFCD00778301 | |
tris 4-aminophenyl amine, 4,4',4-triaminotriphenylamine, n1,n1-bis 4-aminophenyl benzene-1,4-diamine, 1,4-benzenediamine, n,n-bis 4-aminophenyl, n,n-bis 4-aminophenyl-1,4-benzenediamine, 1,4-benzenediamine, n1,n1-bis 4-aminophenyl, 1-n,1-n-bis 4-aminophenyl benzene-1,4-diamine, pubchem22505, tri 4-aminophenyl amine, tris 4-aminophenyl-amine | |
4-N,4-N-bis(4-aminophenyl)benzene-1,4-diamine |
Specifications
5981-09-9 | |
C18H18N4 | |
2812659 | |
Slightly soluble in water. | |
C1=CC(=CC=C1N)N(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N | |
290.37 | |
290.38 | |
Solid |
230°C | |
MFCD00778301 | |
tris 4-aminophenyl amine, 4,4',4-triaminotriphenylamine, n1,n1-bis 4-aminophenyl benzene-1,4-diamine, 1,4-benzenediamine, n,n-bis 4-aminophenyl, n,n-bis 4-aminophenyl-1,4-benzenediamine, 1,4-benzenediamine, n1,n1-bis 4-aminophenyl, 1-n,1-n-bis 4-aminophenyl benzene-1,4-diamine, pubchem22505, tri 4-aminophenyl amine, tris 4-aminophenyl-amine | |
SNLFYGIUTYKKOE-UHFFFAOYSA-N | |
4-N,4-N-bis(4-aminophenyl)benzene-1,4-diamine | |
80083 | |
97% | |
Tris(4-aminophenyl)amine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 227-791-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.