missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-aminophenyl)amine, 97%, Thermo Scientific Chemicals

Product Code. 15460959
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5981-09-9
Molecular Formula C18H18N4
Molecular Weight (g/mol) 290.37
MDL Number MFCD00778301
InChI Key SNLFYGIUTYKKOE-UHFFFAOYSA-N
Synonym tris 4-aminophenyl amine, 4,4',4-triaminotriphenylamine, n1,n1-bis 4-aminophenyl benzene-1,4-diamine, 1,4-benzenediamine, n,n-bis 4-aminophenyl, n,n-bis 4-aminophenyl-1,4-benzenediamine, 1,4-benzenediamine, n1,n1-bis 4-aminophenyl, 1-n,1-n-bis 4-aminophenyl benzene-1,4-diamine, pubchem22505, tri 4-aminophenyl amine, tris 4-aminophenyl-amine
PubChem CID 80083
IUPAC Name 4-N,4-N-bis(4-aminophenyl)benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N

Specifications

Melting Point 230°C
Quantity 1 g
Beilstein 2812659
Solubility Information Slightly soluble in water.
Formula Weight 290.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Tris(4-aminophenyl)amine
compliance-icons
Product Identifier
  • Tris(4-aminophenyl)amine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Tris(4-aminophenyl)amine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.