missing translation for 'onlineSavingsMsg'
Learn More

TRIM44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen TRIM44
Concentration 0.27 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16858643
View Documents
Proteintech
11511-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16848643
View Documents
Proteintech
11511-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This is a rabbit polyclonal antibody raised against full length of human TRIM44.

May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.
Specifications

Specifications

TRIM44
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
Q6QA27, Q96DX7, Q9QXA7
362172, 54765, 80985
TRIM44 Fusion Protein Ag2070
Primary
-20°C
TRIM44
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
DIPB, HSA249128, MC7, Protein DIPB, TRIM44, tripartite motif containing 44
Trim44
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications