Learn More
TRIM44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11511-1-AP-20UL
 View more versions of this product
Product Code. 16858643
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This is a rabbit polyclonal antibody raised against full length of human TRIM44.
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.
Specifications
Specifications
TRIM44
Polyclonal
Unconjugated
TRIM44
DIPB, HSA249128, MC7, Protein DIPB, TRIM44, tripartite motif containing 44
Rabbit
Antigen Affinity Chromatography
RUO
362172, 54765, 80985
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6QA27, Q96DX7, Q9QXA7
Trim44
TRIM44 Fusion Protein Ag2070
20 μL
Primary
Human, Mouse, Rat