Learn More
(Trifluoromethyl)trimethylsilane, 0.5M soln. in THF, Thermo Scientific Chemicals
25.30 € - 218.00 €
Chemical Identifiers
CAS | 81290-20-2 |
---|---|
Molecular Formula | C5H11F3Si |
Molecular Weight (g/mol) | 156.22 |
MDL Number | MFCD00145454 |
InChI Key | MTYSDPUZDZURPP-UHFFFAOYSA-N |
Synonym | trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent |
PubChem CID | 552549 |
IUPAC Name | trimethyl(trifluoromethyl)silane |
SMILES | CC(C)(C)[SiH2]C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15497815
|
Thermo Scientific Alfa Aesar
H31520.AA |
5 mL |
25.30 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407825
|
Thermo Scientific Alfa Aesar
H31520.AC |
25 mL |
78.60 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15417825
|
Thermo Scientific Alfa Aesar
H31520.AE |
100 mL |
218.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA reagent used in nucleophilic addition of the trifluoromethyl group to aldehydes and ketones.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/ moisture.
Chemical Identifiers
81290-20-2 | |
156.22 | |
MTYSDPUZDZURPP-UHFFFAOYSA-N | |
552549 | |
CC(C)(C)[SiH2]C(F)(F)F |
C5H11F3Si | |
MFCD00145454 | |
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent | |
trimethyl(trifluoromethyl)silane |
Specifications
40°C | |
C5H11F3Si | |
MFCD00145454 | |
17°C (63°F) | |
Hydrolyzes in water. | |
CC(C)(C)[SiH2]C(F)(F)F | |
156.22 | |
142.2 | |
Moisture sensitive | |
(Trifluoromethyl)trimethylsilane |
81290-20-2 | |
CF3Si(CH3)3 | |
UN1993 | |
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent | |
MTYSDPUZDZURPP-UHFFFAOYSA-N | |
trimethyl(trifluoromethyl)silane | |
552549 | |
0.5M soln. in THF | |
0.895 |
Safety and Handling
GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
H225-H302-H319-H335-H336-H351
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only