missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethyl)trimethylsilane, 0.5M soln. in THF, Thermo Scientific Chemicals

Product Code. 15417825
Click to view available options
Quantity:
5 mL
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reagent used in nucleophilic addition of the trifluoromethyl group to aldehydes and ketones.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81290-20-2
Molecular Formula C5H11F3Si
Molecular Weight (g/mol) 156.22
MDL Number MFCD00145454
InChI Key MTYSDPUZDZURPP-UHFFFAOYSA-N
Synonym trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
PubChem CID 552549
IUPAC Name trimethyl(trifluoromethyl)silane
SMILES CC(C)(C)[SiH2]C(F)(F)F

Specifications

Boiling Point 40°C
Quantity 100 mL
Linear Formula CF3Si(CH3)3
UN Number UN1993
Flash Point 17°C (63°F)
Solubility Information Hydrolyzes in water.
Formula Weight 142.2
Concentration or Composition (by Analyte or Components) 0.5M soln. in THF
Sensitivity Moisture sensitive
Density 0.895
Chemical Name or Material (Trifluoromethyl)trimethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'