missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin bromide, 95%, Thermo Scientific™

32.90 € - 320.00 €

Chemical Identifiers

CAS 1461-23-0
Molecular Formula C12H27BrSn
Molecular Weight (g/mol) 369.962
MDL Number MFCD00010660
InChI Key FVRKTAOFDKFAMI-UHFFFAOYSA-M
Synonym tributyltin bromide, tri-n-butyltin bromide, bromotributyltin, stannane, tributylbromo, tri-n-butylbromotin, tributyltinbromide, bu3snbr, bromo tributyl stannane, acmc-20ajf5, tri-n-butyl tin bromide
PubChem CID 305767
IUPAC Name bromo(tributyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11334869
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071185.03
1 g
32.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
11324869
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071185.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11314869
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071185.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1461-23-0
369.962
FVRKTAOFDKFAMI-UHFFFAOYSA-M
305767
CCCC[Sn](CCCC)(CCCC)Br
C12H27BrSn
MFCD00010660
tributyltin bromide, tri-n-butyltin bromide, bromotributyltin, stannane, tributylbromo, tri-n-butylbromotin, tributyltinbromide, bu3snbr, bromo tributyl stannane, acmc-20ajf5, tri-n-butyl tin bromide
bromo(tributyl)stannane

Specifications

1461-23-0
163°C (12 mmHg)
C12H27BrSn
MFCD00010660
Moisture sensitive
FVRKTAOFDKFAMI-UHFFFAOYSA-M
bromo(tributyl)stannane
305767
95%
Tri-n-butyltin bromide
1.338 g/mL
>110°C (230°F)
1.507
UN2788
tributyltin bromide, tri-n-butyltin bromide, bromotributyltin, stannane, tributylbromo, tri-n-butylbromotin, tributyltinbromide, bu3snbr, bromo tributyl stannane, acmc-20ajf5, tri-n-butyl tin bromide
CCCC[Sn](CCCC)(CCCC)Br
369.962
369.96
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 215-959-2

missing translation for 'rtecsNumber' : WH6735000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications