missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin bromide, 95%, Thermo Scientific™

Product Code. 11314869
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-23-0
Molecular Formula C12H27BrSn
Molecular Weight (g/mol) 369.962
MDL Number MFCD00010660
InChI Key FVRKTAOFDKFAMI-UHFFFAOYSA-M
Synonym tributyltin bromide, tri-n-butyltin bromide, bromotributyltin, stannane, tributylbromo, tri-n-butylbromotin, tributyltinbromide, bu3snbr, bromo tributyl stannane, acmc-20ajf5, tri-n-butyl tin bromide
PubChem CID 305767
IUPAC Name bromo(tributyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)Br

Specifications

Density 1.338 g/mL
Boiling Point 163°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.507
Quantity 25 g
UN Number UN2788
Sensitivity Moisture sensitive
Formula Weight 369.96
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'