missing translation for 'onlineSavingsMsg'
Learn More

TMEM97 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen TMEM97
Concentration 300 μg/mL
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16844905
View Documents Promotion Details
Proteintech
26444-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16834905
View Documents Promotion Details
Proteintech
26444-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The TMEM97 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TMEM97. This antibody recognizes human, mouse antigen. The TMEM97 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

TMEM97
Western Blot, Immunohistochemistry
Unconjugated
Human, Mouse
IgG
300 μg/mL
Polyclonal
Rabbit
27346
Primary
Videos
SDS
Documents

Documents

Product Certifications