Learn More
TMEM97 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26444-1-AP-150UL
 View more versions of this product
Product Code. 16834905
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The TMEM97 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TMEM97. This antibody recognizes human, mouse antigen. The TMEM97 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TMEM97
Polyclonal
Unconjugated
150 μL
27346
IgG
Western Blot, Immunohistochemistry
300 μg/mL
Rabbit
Primary
Human, Mouse