missing translation for 'onlineSavingsMsg'
Learn More

TMEM70 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen TMEM70
Concentration 0.26 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16859924
View Documents Promotion Details
Proteintech
20388-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16849924
View Documents Promotion Details
Proteintech
20388-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TMEM70 is a recently identified mitochondrial protein that is thought to play a role in the biogenesis of the ATP synthase in higher eukaryotes. Mutations in this gene result in early neonatal onset of hypotonia, hypertrophic cardiomyopathy, lactic acidos and 3-methylglutaconic aciduria (3-MGC-uria), and usually cause death within the first six weeks of life, although some patients survive much longer. Little is known of the role of TMEM70, but it is conserved across multicellular eukaryotic organisms. It contains a conserved DUF1301 domain and two putative transmembrane regions.
Specifications

Specifications

TMEM70
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
Q9BUB7
54968
TMEM70 Fusion Protein Ag14226
Primary
-20°C
TMEM70
0.26 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
TMEM70, transmembrane protein 70
TMEM70
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates