Learn More
TMEM70 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20388-1-AP-20UL
 View more versions of this product
Product Code. 16859924
  • 145.00 € / 20µL
Explore more special offers
Description
Description
TMEM70 is a recently identified mitochondrial protein that is thought to play a role in the biogenesis of the ATP synthase in higher eukaryotes. Mutations in this gene result in early neonatal onset of hypotonia, hypertrophic cardiomyopathy, lactic acidos and 3-methylglutaconic aciduria (3-MGC-uria), and usually cause death within the first six weeks of life, although some patients survive much longer. Little is known of the role of TMEM70, but it is conserved across multicellular eukaryotic organisms. It contains a conserved DUF1301 domain and two putative transmembrane regions.
Specifications
Specifications
TMEM70
Polyclonal
Unconjugated
TMEM70
TMEM70, transmembrane protein 70
Rabbit
Antigen Affinity Chromatography
RUO
54968
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BUB7
TMEM70
TMEM70 Fusion Protein Ag14226
20 μL