missing translation for 'onlineSavingsMsg'
Learn More

TMEM173/STING Mouse anti-Human, Rat, Clone: 1F1E1, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen TMEM173/STING
Clone 1F1E1
Concentration 1 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16868785
View Documents Promotion Details
Proteintech
66680-1-IG-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16858785
View Documents Promotion Details
Proteintech
66680-1-IG-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

STING is a 379 amino acid containing signaling protein that acts as an important component of innate immune signaling. This signaling protein is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon (IFN-alpha and IFN-beta) and exert a potent anti-viral state following expression. STING may have translocon function, the translocon possibly being able to influence the induction of type I interferons and is also involved in transduction of apoptotic signals via its association with the major histocompatibility complex class II (MHC-II) thus facilitating the detection of intracellular viral RNA species as well as B-form DNA and mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. Generally seen in endoplasmic reticulum membrane, cell membrane and mitochondrion outer membrane, it is ubiquitously expressed in most of tissues.
Specifications

Specifications

TMEM173/STING
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, MPYS, STING, TMEM173, transmembrane protein 173
TMEM173
IgG2b
Protein A
Antibody
1F1E1
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Rat
F1M391, Q86WV6
340061, 498840
TMEM173/STING Fusion Protein Ag20363
Primary
-20°C
TMEM173
Videos
SDS
Documents

Documents

Product Certifications