missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM173/STING Mouse anti-Human, Rat, Clone: 1F1E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66680-1-IG-150UL
This item is not returnable.
View return policy
Description
STING is a 379 amino acid containing signaling protein that acts as an important component of innate immune signaling. This signaling protein is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon (IFN-alpha and IFN-beta) and exert a potent anti-viral state following expression. STING may have translocon function, the translocon possibly being able to influence the induction of type I interferons and is also involved in transduction of apoptotic signals via its association with the major histocompatibility complex class II (MHC-II) thus facilitating the detection of intracellular viral RNA species as well as B-form DNA and mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. Generally seen in endoplasmic reticulum membrane, cell membrane and mitochondrion outer membrane, it is ubiquitously expressed in most of tissues.Specifications
TMEM173/STING | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
F1M391, Q86WV6 | |
TMEM173 | |
TMEM173/STING Fusion Protein Ag20363 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1F1E1 | |
Unconjugated | |
TMEM173 | |
FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, MPYS, STING, TMEM173, transmembrane protein 173 | |
Mouse | |
Protein A | |
RUO | |
340061, 498840 | |
-20°C | |
Liquid |