missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM127 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | TMEM127 |
---|---|
Concentration | 0.47 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16802215
|
Proteintech
23142-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16892205
|
Proteintech
23142-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM127 controls cell proliferation acting as a negative regulator of TOR signaling pathway mediated by mTORC1. May act as a tumor suppressor.Specifications
TMEM127 | |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O75204, Q8BGP5 | |
55654, 69470 | |
TMEM127 Fusion Protein Ag19472 | |
Primary | |
-20°C | |
TMEM127 |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TMEM127, transmembrane protein 127 | |
TMEM127 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title