missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM127 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23142-1-AP-150UL
This item is not returnable.
View return policy
Description
TMEM127 controls cell proliferation acting as a negative regulator of TOR signaling pathway mediated by mTORC1. May act as a tumor suppressor.Specifications
TMEM127 | |
Polyclonal | |
Unconjugated | |
TMEM127 | |
TMEM127, transmembrane protein 127 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55654, 69470 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75204, Q8BGP5 | |
TMEM127 | |
TMEM127 Fusion Protein Ag19472 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction