Learn More
Masterflex™ Thread to Compression Adapters
Compression fittings are an excellent choice for connecting rigid tubing systems
68.15 € - 128.00 €
Specifications
Description | Compression x NPT(M) Connection |
---|---|
Autoclavable | Not Autoclavable |
Material | Polypropylene |
Packaging Quantity | 10 Fittings |
Tubing Size | 3/4 in. |
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Available in packs of ten
Specifications
Compression x NPT(M) Connection | |
Polypropylene | |
3/4 in. | |
Ethylene Oxide |
Not Autoclavable | |
10 Fittings | |
3/4 in. | |
Straight |