Learn More
Tetraphenyltin, 97%, Thermo Scientific™
57.05 € - 162.00 €
Chemical Identifiers
CAS | 595-90-4 |
---|---|
Molecular Formula | C24H20Sn |
Molecular Weight (g/mol) | 427.13 |
MDL Number | MFCD00003006 |
InChI Key | CRHIAMBJMSSNNM-UHFFFAOYSA-N |
Synonym | tetraphenyltin, stannane, tetraphenyl, tetraphenyl tin, ccris 6326, tin, tetraphenyl-,, stannane,tetraphenyl, acmc-209mem |
PubChem CID | 61146 |
IUPAC Name | tetraphenylstannane |
SMILES | C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15433187
|
Thermo Scientific Alfa Aesar
A19132.09 |
10 g |
57.05 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11385249
|
Thermo Scientific Alfa Aesar
A19132.18 |
50 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
595-90-4 | |
427.13 | |
CRHIAMBJMSSNNM-UHFFFAOYSA-N | |
61146 | |
C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C24H20Sn | |
MFCD00003006 | |
tetraphenyltin, stannane, tetraphenyl, tetraphenyl tin, ccris 6326, tin, tetraphenyl-,, stannane,tetraphenyl, acmc-209mem | |
tetraphenylstannane |
Specifications
595-90-4 | |
>420°C | |
Characteristic | |
MFCD00003006 | |
Light sensitive | |
CRHIAMBJMSSNNM-UHFFFAOYSA-N | |
tetraphenylstannane | |
61146 | |
97% |
224°C to 227°C | |
>110°C (230°F) | |
C24H20Sn | |
UN3146 | |
tetraphenyltin, stannane, tetraphenyl, tetraphenyl tin, ccris 6326, tin, tetraphenyl-,, stannane,tetraphenyl, acmc-209mem | |
C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
427.13 | |
427.11 | |
Tetraphenyltin |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 209-872-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures