missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenyltin, 97%, Thermo Scientific™

Product Code. 15433187
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-90-4
Molecular Formula C24H20Sn
Molecular Weight (g/mol) 427.13
MDL Number MFCD00003006
InChI Key CRHIAMBJMSSNNM-UHFFFAOYSA-N
Synonym tetraphenyltin, stannane, tetraphenyl, tetraphenyl tin, ccris 6326, tin, tetraphenyl-,, stannane,tetraphenyl, acmc-209mem
PubChem CID 61146
IUPAC Name tetraphenylstannane
SMILES C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C to 227°C
Boiling Point >420°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 10 g
UN Number UN3146
Sensitivity Light sensitive
Formula Weight 427.11
Percent Purity 97%
Chemical Name or Material Tetraphenyltin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'