Learn More
Tetra-n-propoxysilane, 97%, Thermo Scientific Chemicals
40.90 € - 117.59 €
Chemical Identifiers
CAS | 682-01-9 |
---|---|
Molecular Formula | C12H28O4Si |
Molecular Weight (g/mol) | 264.437 |
MDL Number | MFCD00009364 |
InChI Key | ZQZCOBSUOFHDEE-UHFFFAOYSA-N |
Synonym | tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane |
PubChem CID | 69627 |
IUPAC Name | tetrapropyl silicate |
SMILES | CCCO[Si](OCCC)(OCCC)OCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389295
|
Thermo Scientific Alfa Aesar
L16644.14 |
25 g |
40.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399295
|
Thermo Scientific Alfa Aesar
L16644.22 |
100 g |
117.59 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
682-01-9 | |
264.437 | |
ZQZCOBSUOFHDEE-UHFFFAOYSA-N | |
69627 | |
CCCO[Si](OCCC)(OCCC)OCCC |
C12H28O4Si | |
MFCD00009364 | |
tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane | |
tetrapropyl silicate |
Specifications
224°C to 225°C (94°C (5mmHg)) | |
C12H28O4Si | |
MFCD00009364 | |
95°C (203°F) | |
ZQZCOBSUOFHDEE-UHFFFAOYSA-N | |
tetrapropyl silicate | |
69627 | |
97% | |
Moisture sensitive | |
Tetra-n-propoxysilane |
682-01-9 | |
(CH3CH2CH2O)4Si | |
1706115 | |
tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane | |
CCCO[Si](OCCC)(OCCC)OCCC | |
264.437 | |
264.44 | |
1.401 | |
0.916 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 211-659-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.