missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propoxysilane, 97%, Thermo Scientific Chemicals

Product Code. 11389295
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 682-01-9
Molecular Formula C12H28O4Si
Molecular Weight (g/mol) 264.437
MDL Number MFCD00009364
InChI Key ZQZCOBSUOFHDEE-UHFFFAOYSA-N
Synonym tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane
PubChem CID 69627
IUPAC Name tetrapropyl silicate
SMILES CCCO[Si](OCCC)(OCCC)OCCC

Specifications

Boiling Point 224°C to 225°C (94°C (5mmHg))
Quantity 25 g
Linear Formula (CH3CH2CH2O)4Si
Beilstein 1706115
Flash Point 95°C (203°F)
Formula Weight 264.44
Percent Purity 97%
Refractive Index 1.401
Sensitivity Moisture sensitive
Density 0.916
Chemical Name or Material Tetra-n-propoxysilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.