Learn More
Dostmann Electronic™ Immersion Temperature Probe
Dostmann Electronic™ Immersion Temperature Probe is useful for temperature measurements in liquids and powdered materials.
Brand:  Dostmann Electronic™ 6010-0575
 View more versions of this product
Product Code. 11836850
  • 43.30 € / Each
Explore more special offers
Description
Description
  • Type K according to EN 60584-1
  • NiCr-Ni
  • Mineral-insulated
Specifications
Specifications
1 mm
No handle, or cable
Temperature Probe
-200°C to +1100°C
For Measuring In Liquid And Powdered Materials
130 mm
Flexible probe
P410 / TC301 / TC 309 / Mini-K