Learn More
Dostmann Electronic™ Immersion Temperature Probe
Dostmann Electronic™ Immersion Temperature Probe is useful for temperature measurements in liquids and powdered materials.
Brand:  Dostmann Electronic™ 6010-0503
 View more versions of this product
Product Code. 10661366
  • 102.00 € / Each
Explore more special offers
Description
Description
  • Type K according to EN 60584-1
  • NiCr-Ni
  • Mineral-insulated
Specifications
Specifications
3 mm
Handle and cable
Temperature Probe
-100°C to +1100°C
For Measuring In Liquid And Powdered Materials
300 mm
Non-flexible probe
P410 / TC301 / TC 309 / Mini-K