Learn More
Usbeck Laborgeraete™ Teclu Burner
Conduct experimentation at hotter temperatures with the Usbeck Laborgeraete™ Teclu Burner. It is available for use with natural gas and propane, with achievable temperatures up to 1600°C.
Brand: Usbeck Laborgeraete™ 1221
Alert:
Burners according to DIN 30665 cannot be switched off completely. A small flame remains burning even if needle valve or stop cock are turned off. The gas flow shall be disconnected by means of the central stopcock in the laboratory.
Description
- Air regulation
- Needle valve
- Variant of Bunsen burner
Specifications
Teclu Burner | |
17 mm (Head) | |
155 mm | |
1 Ea. |
Propane | |
14 mm | |
1530°C | |
260 g |