missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF3 Mouse anti-Human, Unconjugated, Clone: 1D12E5, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | TCF3 |
---|---|
Clone | 1D12E5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16809605
|
Proteintech
67140-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16899595
|
Proteintech
67140-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TCF3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCF3. This antibody recognizes human antigen. The TCF3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
TCF3 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
6929 | |
Primary |
1D12E5 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG2b |