Learn More
TCF3 Mouse anti-Human, Unconjugated, Clone: 1D12E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67140-1-IG-20UL
This item is not returnable.
View return policy
Description
The TCF3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCF3. This antibody recognizes human antigen. The TCF3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
TCF3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
1D12E5 | |
Unconjugated | |
20 μL | |
6929 | |
IgG2b |