missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNJ2BP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | SYNJ2BP |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894934
|
Proteintech
15666-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884934
|
Proteintech
15666-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Regulates endocytosis of activin type 2 receptor kinases through the Ral/RALBP1-dependent pathway and may be involved in suppression of activin-induced signal transduction.Specifications
SYNJ2BP | |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P57105, Q9D6K5 | |
24071, 55333 | |
SYNJ2BP Fusion Protein Ag8130 | |
Primary | |
-20°C | |
SYNJ2BP |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARIP2, OMP25, synaptojanin 2 binding protein, SYNJ2BP | |
SYNJ2BP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |