missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNJ2BP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15666-1-AP-20UL
This item is not returnable.
View return policy
Description
Regulates endocytosis of activin type 2 receptor kinases through the Ral/RALBP1-dependent pathway and may be involved in suppression of activin-induced signal transduction.Specifications
SYNJ2BP | |
Polyclonal | |
Unconjugated | |
SYNJ2BP | |
ARIP2, OMP25, synaptojanin 2 binding protein, SYNJ2BP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24071, 55333 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P57105, Q9D6K5 | |
SYNJ2BP | |
SYNJ2BP Fusion Protein Ag8130 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |