Learn More
IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity FPLC column
Strep-Tactin™ 4Flow™ high capacity FPLC column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand:  IBA Lifesciences 2-1258-001
 View more versions of this product
Product Code. 30117607
  • 421.00 € / 5mL
Explore more special offers
Description
Description
  • Strep-Tactin™ 4Flow™ high capacity FPLC columns are designed for purifying Strep-tag™II and Twin-Strep-tag™ proteins via affinity chromatography in automated protein purification with HPLC/FPLC devices such as Äkta™ systems (Cytiva).
  • Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
  • 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
  • Multiple regeneration cycles possible without loss of binding capacity
  • Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
  • Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock
Specifications
Specifications
20 mg/mL resin
4% agarose, crosslinked
Resin Slurry
Tetrameric protein
Column
Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
FPLC cloumn
5 mL
50 to 150 μm
5 mL
Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Affinity chromatography equipment
Agarose
Pre-packed gravity flow column