Learn More
SP Bel-Art™ Stirring Bars Spinbar™
Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
9.10 € - 39.48 €
Specifications
Shape Octagon
View More Specs
Products 10
Product Code Brand Length (Metric) Diameter (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Price Quantity & Availability  
16438485
View Documents
SP Bel-Art™
F37110-1308
13 mm 8 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16448485
View Documents
SP Bel-Art™
F37110-1608
16 mm 8 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16458485
View Documents
SP Bel-Art™
F37110-2208
22 mm 8 mm
27.95 €
Pack of 10
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
16468485
View Documents
SP Bel-Art™
F37110-2508
25 mm 8 mm
39.48 €
Pack of 10
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
16478485
View Documents
SP Bel-Art™
F37110-2908
29 mm 8 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16488485
View Documents
SP Bel-Art™
F37110-3808
38 mm 8 mm
36.75 €
Pack of 10
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
16498485
View Documents
SP Bel-Art™
F37110-3810
38 mm 10 mm
39.20 €
Pack of 10
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
16408495
View Documents
SP Bel-Art™
F37110-5108
51 mm 8 mm
20.60 €
Pack of 5
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
16418495
View Documents
SP Bel-Art™
F37110-5110
51 mm 10 mm
33.00 €
Pack of 5
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
16428495
View Documents
SP Bel-Art™
F37110-7613
76 mm 13 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • With pivot ring for better mixing conditions
  • Alnico magnet encapsulated with PTFE™ brand PTFE coating
  • FDA Grade, USP Class VI
  • Good chemical compatibility
  • Autoclavable
Specifications
Specifications
Octagon