Learn More
SP Bel-Art™ Stirring Bars Spinbar™
Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
Brand:  SP Bel-Art™ F37110-2508
 View more versions of this product
Product Code. 16468485
  • 39.48 € / Pack of 10
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With pivot ring for better mixing conditions
  • Alnico magnet encapsulated with PTFE™ brand PTFE coating
  • FDA Grade, USP Class VI
  • Good chemical compatibility
  • Autoclavable
Specifications
Specifications
8 mm
25 mm
10/Pk.
Octagon