missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StarGate pESG-IBA Vectors for Mammalian Expression

Designed for high-level constitutive expression of recombinant proteins in a wide range of Mammalian host cells through the CMV promoter, with replication in cell lines that are latently infected with SV40 large T antigen.

Brand:  IBA Lifesciences 5-4465-001

 View more versions of this product

Product Code. 15683788

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

pESG-IBA mammalian expression vectors

pESG-IBA is a small expression vector recommended for transient expression of proteins in cell lines providing SV40 large T-antigen for extrachromosomal replication with SV40 ori.

  • NeoR for generation of stable cells lines.
  • Cytomegalovirus immediate-early (CMV) promoter: high level constitutive expression
  • SV40 ori: origin of replication for extrachromosomal replication driven by SV40 large T-antigen (as provided by HEK293T or COS-1, -3, -7 cells)
  • ColE1 + Amp: for propagation in E. coli
  • NeoR: neomycin resistance gene to generate stable cell lines
  • BM40: secretion signal to transfer the expressed protein into the medium (not for all vectors available)

Please note that cloning into pESG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pESG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pESG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.

Specifications

Specifications

Liquid
pESG-IBA 65
Neomycin resistance gene
Human CMV Immediate-early
20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
FLAG-tag for N-terminal
5 μg
Cloning Genes of Interest for Use in Functional Expression, Purification, Detection, Immobilization or Separation of Recombinant Proteins
StarGate™ System, Mammalian Host Cells
SV40 large T antigen (e.g. COS1 or COS7)
8
Acceptor Vector for mammalian expression
2°C to 8°C for frequent usage, -20°C for long-term storage
250 ng/μL
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.