missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Stainless Steel Beakers With Lid And Handle
Nickel Electro™ Stainless Steel Beakers With Lid And Handle to complete their seamless, crevice free exterior.
59.85 € - 156.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12767856
|
Nickel Electro™
11001 |
1000 mL |
59.85 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
12777856
|
Nickel Electro™
11002 |
2000 mL |
108.00 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
12787856
|
Nickel Electro™
11003 |
3000 mL |
107.00 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
12797856
|
Nickel Electro™
11004 |
5000 mL |
156.00 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
Description
- Stainless steel
- Includes lid
- Seamless and crevice free exterior
- Non-magnetic
- Flat base for stability
- All surfaces are non-reacting and offer zero absorption
- All surfaces are highly polished making cleaning easier
- Can be sterilized by autoclaving
Specifications
Stainless Steel | |
Lid, Handle | |
Beaker |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title