missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%
37.20 € - 680.80 €
Chemical Identifiers
CAS | 10102-17-7 |
---|---|
Molecular Formula | H10Na2O8S2 |
Molecular Weight (g/mol) | 248.172 |
InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
PubChem CID | 61475 |
ChEBI | CHEBI:32150 |
IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
SMILES | O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15683450
|
Honeywell-Fluka
31459-500G |
500 g | N/A | ||||||
15673450
|
Honeywell-Fluka
31459-1KG |
1 kg |
61.50 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15603460
|
Honeywell-Fluka
31459-5KG |
5 kg | N/A | ||||||
15613460
|
Honeywell-Fluka
31459-6X1KG |
6 x 1 kg |
287.26 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15693450
|
Honeywell-Fluka
31459-50KG |
50 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10102-17-7 | |
248.172 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
CHEBI:32150 | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
H10Na2O8S2 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
Specifications
10102-17-7 | |
Na2S2O3 · 5H2O | |
NONH for all modes of transport | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
61475 | |
248.18g/mol | |
ACS/puriss. p.a. |
H10Na2O8S2 | |
MFCD00149186 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
248.172 | |
CHEBI:32150 | |
≥99.5% | |
Sodium Thiosulfate Pentahydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
missing translation for 'einecsNumber' : 231-867-5