missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Product Code. 15603460
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Quantity 5 kg
Linear Formula Na2S2O3 · 5H2O
MDL Number MFCD00149186
UN Number NONH for all modes of transport
Formula Weight 248.18g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Thiosulfate Pentahydrate, Reag. Ph. Eur. reag. ISO
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'