missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic, puriss p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)
39.98 € - 389.32 €
Chemical Identifiers
CAS | 7558-79-4 |
---|---|
Molecular Formula | HNa2O4P |
Molecular Weight (g/mol) | 141.96 |
MDL Number | MFCD00003496 |
InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
Synonym | disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt |
PubChem CID | 24203 |
ChEBI | CHEBI:34683 |
IUPAC Name | disodium;hydrogen phosphate |
SMILES | [Na+].[Na+].OP([O-])([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15616810
|
Honeywell-Fluka
71640-250G |
250 g |
39.98 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15606810
|
Honeywell-Fluka
71640-1KG |
1 kg |
124.02 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15626810
|
Honeywell-Fluka
71640-5KG |
5 kg |
389.32 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7558-79-4 | |
141.96 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
24203 | |
disodium;hydrogen phosphate |
HNa2O4P | |
MFCD00003496 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
CHEBI:34683 | |
[Na+].[Na+].OP([O-])([O-])=O |
Specifications
7558-79-4 | |
Na2HPO4 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
[Na+].[Na+].OP([O-])([O-])=O | |
141.96 | |
CHEBI:34683 | |
≥99% (T) | |
Sodium phosphate dibasic |
HNa2O4P | |
MFCD00003496 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium;hydrogen phosphate | |
24203 | |
141.96g/mol | |
ACS/puriss. p.a. |
Safety and Handling
missing translation for 'rtecsNumber' : WC4500000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title