missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic, puriss p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 1 kg |
Linear Formula | Na2HPO4 |
Formula Weight | 141.96g/mol |
Percent Purity | ≥99% (T) |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Sodium phosphate dibasic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction