missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, meets analytical specification of Ph.Eur
38.35 € - 269.00 €
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11984051
|
Fisher Chemical
S/3161/60 |
1 kg | Plastic powder jar |
38.35 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11994051
|
Fisher Chemical
S/3161/63 |
3 kg | Plastic powder jar | N/A | |||||
11994929
|
Fisher Chemical
S/3161/68 |
10 kg | Plastic keg |
179.00 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964909
|
Fisher Chemical
S/3161/70 |
25 kg | Plastic keg |
269.00 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 |
Specifications
7647-14-5 | |
MFCD00003477 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium;chloride | |
5234 |
ClNa | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title